Pyronin G ≥99%, certified (C.I. 45005 - C.N. DcPy-2)

Supplier: Electron Microscopy Sciences
Danger

19560
100504-198EA 129.79 USD
100504-198
Pyronin G ≥99%, certified (C.I. 45005 - C.N. DcPy-2)
Pyronine G

Cudder's method combined gram/pappenheim stain for gonorrheal pus. Kurnick's method used with methyl green which stains liver cells. For staining protein in the diazosulfanilic acid technique; a substitute for azur A. can be used for pyronin B.


Solubility : 9.0% Water; 0.5% Alcohol; 1.85% Cellosolve; 4.35% Glycol; 0.0% Xylene

Formula: C₁₇H₁₉ClN₂O
Melting Pt: 250…260 °C (482…500 °F)
Density: 0.64 g/cm³ (5.341 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00011725
CAS Number: 92-32-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95f43bdeb3effa64fce82578f8bdc959dc386b95&vid=95f43bdeb3effa64fce82578f8bdc959dc386b95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR