Phenol red

Supplier: Electron Microscopy Sciences
Warning

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

19327
100504-184EA 34.68 USD
100504-184
Phenol red
Phenol red

A standard indicator for preparing buffers, pH range 6.4 to 8.2.

Formula: C₁₉H₁₄O₅S
Storage Temperature: Ambient
MDL Number: MFCD00003552
CAS Number: 143-74-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88733c951d1d3af75677fb4e7d3e001383bca47a&vid=88733c951d1d3af75677fb4e7d3e001383bca47a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR