Orange G, certified (C.I. 16230)

Supplier: Electron Microscopy Sciences

19070
100504-126EA 62.9 USD
100504-126
Orange G, certified (C.I. 16230)
Orange G

Kornhauser's quad stain for most elementary structures of tissues. Stain Tech., 18, 985 (1943) Mallory's tissue stain for connective tissue. Mallory, J. Exper. Med., 5,15 (1900). Used with safranin and tannic acid for showing shoot apex in plant tissue. Stain Tech., 18, 105 (1943).

Formula: C₁₆H₁₀N₂Na₂O₇S₂
Storage Temperature: Ambient
MDL Number: MFCD00012457
CAS Number: 1936-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73229b0c0d6f792eee1d1b09a0343fe3caf4a33f&vid=73229b0c0d6f792eee1d1b09a0343fe3caf4a33f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Dye Content Approx. 60%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR