Methyl cellulose, 400 cPs USP, NF
Supplier: Electron Microscopy Sciences
|
Warning
|
18560
100504-102EA
99.23
USD
100504-102
Methyl cellulose, 400 cPs USP, NF
Methyl cellulose
(Methyl Alcohol)
CH3OH F.W. 32.04 CAS 67-56-1
Assay: 99.8%
Specific Gravity : 0.79
A.C.S.
Storage Temperature:
Ambient |
MDL Number:
MFCD00081763 CAS Number: 9004-67-5 |
|
Specification Test Results
Methoxyl (-CH₃) Content | 27.5 - 31.5% |
Medium Viscosity (2% in water, at 20 °C) | 230 - 550 cps |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...