Magnesium chloride hexahydrate ACS

Supplier: Electron Microscopy Sciences

18010
100504-090EA 46.24 USD
100504-090
Magnesium chloride hexahydrate ACS
Magnesium chloride hexahydrate

MgCl2·6H2O F.W. 203.30 Assay: 99.0-102.0%

Formula: MgCl₂·6H₂O
MDL Number: MFCD00149781
CAS Number: 7791-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR