Lead(II) nitrate ≥99.5% ACS

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Lead dinitrate

17900
100504-076EA 28.64 USD
100504-076
Lead(II) nitrate ≥99.5% ACS
Lead (II) nitrate

A metal stain for ultra thin sections.


For complete details please see Sato, T., (1967) J. Electron Micro., 16, 733.

Formula: Pb(NO₃)₂
MW: 331.21 g/mol
Melting Pt: 470 °C (878 °F)
Density: 4.535 g/cm³ (37.845 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
UN: 1469
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd2e7028b4824170d0a1568f338ec6e0fa4a4c4a&vid=dd2e7028b4824170d0a1568f338ec6e0fa4a4c4a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Minimum Assay 99.5%
Insoluble Matters 0.005%
Reaction Not less than pH 3.5
Chloride 0.0005%
Calcium 0.01%
Copper 0.0005%
Iron 0.0005%
Potassium 0.002%
Sodium 0.002%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR