Giemsa's stain, dry, powder, certified by the Biological Stain Commission

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

15942
100503-944EA 68.7 USD
100503-944
Giemsa's stain, dry, powder, certified by the Biological Stain Commission
Giemsa's stain

Thin film stain for differentiation of types of leucocytes; Rickettsiae, bacteria, & inclusion bodies. May-Grunwald/Giemsa for bone marrow stain. Pinkus' acid orcein-giemsa for connective tissue staining

Boiling Pt: 442 °C (828 °F)
Melting Pt: 300 °C (572 °F)
Density: 0.99 g/cm³ (8.262 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db6d5be2be4db0648c5f9bb4ef4b4f41a2897d1f&vid=db6d5be2be4db0648c5f9bb4ef4b4f41a2897d1f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR