Formamide, Ultra High Purity

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Carbamaldehyde

15745 15746 15747
100503-924EA 79 USD
100503-924 100503-926 100503-928
Formamide, Ultra High Purity
Formamide

Glass Distilled. Exceeds ACS Specifications.

Formula: HCONH₂
Boiling Pt: 210.5 °C (411 °F)
Melting Pt: 2.5 °C (37 °F)
Density: 1.1339 g/cm³ (9.462 lbs/gal.)
Flash Pt: 175 °C (347 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007941
CAS Number: 75-12-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8551cdc1f6553f43f7da473306b1937dd6ae3727&vid=8551cdc1f6553f43f7da473306b1937dd6ae3727&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.9%
Copper 0.1 ppm
Iron 0.5 ppm
Lead 0.5 ppm
Specific Gravity 1.128 - 1.132
Zinc 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR