EDTA disodium salt dihydrate ≥99% ACS

Supplier: Electron Microscopy Sciences
Warning

15200
100503-890EA 127.94 USD
100503-890
EDTA disodium salt dihydrate ≥99% ACS
EDTA disodium salt dihydrate

Organic chelating for staining glutaraldehyde fixed tissues, pre-stained with uranyl acetate, followed by lead citrate.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99+%
Heavy Metal (as Pb) Max 0.005%
Insoluble Matters Max. 0.005%
Iron Max. 0.01%
Nitrilotriacetic Acid Max 0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR