Erythrosin B disodium salt, certified (C.I. 45430)

Supplier: Electron Microscopy Sciences
Danger

15010
100503-888EA 75.54 USD
100503-888
Erythrosin B disodium salt, certified (C.I. 45430)
Erythrosin B disodium salt

Used with methylene blue as a plasma stain for nerve cells and use crystal violet for plant tissue.


Solubility : 10.0% Water; 5.0% Alcohol; 7.0% Cellosolve; 8.25% Glycol; 0.0% Xylene

Formula: C₂₀H₆I₄Na₂O₅
Density: 1 g/cm³ (8.345 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00144257
CAS Number: 16423-68-0
UN: 2810
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5d8292b322381160b17eb037bfdf9182889ac58&vid=d5d8292b322381160b17eb037bfdf9182889ac58&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR