Dimethyl sulfoxide ACS

Supplier: Electron Microscopy Sciences
Warning

13390
100503-832EA 375.81 USD
100503-832
Dimethyl sulfoxide ACS
Dimethyl sulphoxide

(CH3)2SO FW 78.13 CAS 67-68-5 A.C.S.

Formula: (CH₃)₂SO
Melting Pt: 18.45 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e173b6e3631cb0532d83d7cb4f07c96baa51026&vid=40b59219d8421bad0f95a7d46f7394c35ae8ece5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, Colorless
Freezing Point Not below 18.0°C
Residue After Evaporation 0.01%
Water 0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR