Acetonedimethyl acetal
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
2,2-Dimethoxypropane, Dimethoxypropane, DMP, Dimethoxy-2,2-propane
Used as a dehydration agent.
Miller, L.L. and Jacks, T.J. (1975). Rapid chemical dehydration of samples for electron microscopic examinations. J. Histochem. Cytochem, 23:107
Thorpe J.R. and Harvey D.M.R. (1979). Optimization and investigation of the use of 2,2-dimethoxypropane as a dehydration agent for plant tissue in TEM. J. Ultrastrut. Res. ,68:186
ry, 76:407
Beckmann H.J. and Dierichs, R. (1982). Lipid extracting properties of 2,2-Dimethoxypropane as revealed by electron microscopy and thin layer chromatography Histochemist
Formula:
C₅H₁₂O₂ Boiling Pt: 83 °C Density: 0.85 g/cm³ (7.093 lbs/gal.) Flash Pt: –10 °C (14 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008479 CAS Number: 77-76-9 UN: 1993 ADR: 3,II |
|
Specification Test Results
Specific Gravity | 0.849 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...