Digitonin, crystals, Reagent Grade

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Digitonin

13260
100503-820EA 768.7 USD
100503-820
Digitonin, crystals, Reagent Grade
Digitonin

An additive which stabilizes cholesterol 0.2% digitonin in buffer added to Glutaraldehyde fixative may help to stabilize and retain the fine structural localization of free cholesterol and cholesterol esters.


Okros, I Histochemie 13-91, 1968; Windaus, A. Hoop Seyler's Z. Physio. Chem. 65:110, 1910. J. Cell Bio., 40, 802, 1969; Temkin, Robert J. (1993); Microscopy Research and Tech. 26:260-271.

Formula: C₅₆H₉₂O₂₉
MW: 1229.312 g/mol
Melting Pt: 230…240 °C (446…464 °F)
Storage Temperature: Ambient
MDL Number: MFCD00077729
CAS Number: 11024-24-1
UN: 3462
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19ff45ec9691dffc79b9d6d2591d127b5f0d38f6&vid=19ff45ec9691dffc79b9d6d2591d127b5f0d38f6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR