Butyl acetate ACS

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate

11900
100503-736EA 64.55 USD
100503-736
Butyl acetate ACS
Butyl acetate

CH3COO(CH2)3CH3 F.W. 116.16 b.p . 127°C Specific Gravity: 0.88

Formula: CH₃COO(CH₂)₃CH₃
MW: 116.16 g/mol
Boiling Pt: 126
Melting Pt: -76
Density: 0.88 g/cm³ (7.344 lbs/gal.)
Flash Pt: 27 °C (81 °F)
MDL Number: MFCD00009445
CAS Number: 123-86-4
UN: 1123
ADR: 3 III
Merck Index: 13,01534
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70affffb1f04f8e702181ef4e02aa199005f8d2e&vid=70affffb1f04f8e702181ef4e02aa199005f8d2e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Specific Gravity 0.88


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR