Bismuth(III) nitrate basic, crystalline

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate

11450
100503-694EA 59.03 USD
100503-694
Bismuth(III) nitrate basic, crystalline
Bismuth (III) nitrate basic

Bismuth staining for light and electron microscopy.

Formula: Bi₅O(OH)₉(NO₃)₄
Density: 4.93 g/cm³ (41.141 lbs/gal.)
MDL Number: MFCD00064844
CAS Number: 1304-85-4
UN: 1477
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR