Basic fuchsin, certified CI 42510, CN DcFb-26

Supplier: Electron Microscopy Sciences
Warning

Synonyms: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline

11260
100503-678EA 64.18 USD
100503-678
Basic fuchsin, certified CI 42510, CN DcFb-26
Basic Fuchsin

Can be used for gram positive/negative bacteria and as a pituitary stain.


Can also be used in the selective staining of nervous tissue for light microscopy following preparation for electron microscopy (Berkowitz et al., J. Histochem. Cytochem. 16:808, 1968.
Can be used as a simple rapid polychrome stain for epoxy-embedded tissue (Sato, T., and Shamoto, M. Stain Technol. 48:223, 1973).


Certifications: Certified by the Biological Stain Commission.


Caution: All Dyes/Stains are for research use only. Not meant for drug, food household use.

Formula: C₂₀H₂₀ClN₃
MW: 337.85 g/mol
Melting Pt: 205 °C (401 °F)
MDL Number: MFCD00012569
CAS Number: 632-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR