Amyl acetate, purified, Glass Distilled

Supplier: Electron Microscopy Sciences
Warning

Synonyms: n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate

10816 10815
100503-662EA 167.12 USD
100503-662 100503-660
Amyl acetate, purified, Glass Distilled
Amyl acetate

Purified, glass distilled amyl acetate is available to meet your laboratory needs.

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 149
Melting Pt: -71
Density: 0.88 g/cm³ (7.344 lbs/gal.)
Flash Pt: 38 °C (100 °F)
MDL Number: MFCD00009500
CAS Number: 628-63-7
UN: 1104
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa32b74ee03ddae6126df9f05771ac4ebe887b46&vid=718009b6e330a5ab836f98bdd1adba048ba763c0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR