Amyl acetate ACS

Supplier: Electron Microscopy Sciences
Warning

Synonyms: n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate

10800 10810
100503-656EA 118.82 USD
100503-656 100503-658
Amyl acetate ACS
Amyl acetate

CH3COOC5H11 FW 130.18 CAS 628-63-7 Boiling Range: 140.0°C-150.0°C Specific Gravity 25° C: 0.870-0.875 A.C.S.

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 149
Melting Pt: -71
Density: 0.8752 g/cm³ (7.304 lbs/gal.)
Flash Pt: 16…25 °C (61…77 °F)
MDL Number: MFCD00009500
CAS Number: 628-63-7
UN: 1104
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa32b74ee03ddae6126df9f05771ac4ebe887b46&vid=718009b6e330a5ab836f98bdd1adba048ba763c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acidity (as CH₃COOH) 0.01%
Non-volatile Matters 0.01%
Specific Gravity at 25 °C 0.870 - 0.875
Water (W/W) 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR