Ammonium heptamolybdate tetrahydrate ≥99%

Supplier: Electron Microscopy Sciences
Warning

10500
100503-650EA 148.89 USD
100503-650
Ammonium heptamolybdate tetrahydrate ≥99%
Ammonium heptamolybdate tetrahydrate

Used in the negative staining of viruses for TEM. Modifications caused by uncoupling agents (Muscatello, U., et al, J. Ultrastruct., Res., 52, 2 (1975)).

Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Melting Pt: 287 °C (549 °F)
Density: 1.782 g/cm³ (14.871 lbs/gal.)
MDL Number: MFCD00167059
CAS Number: 12054-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR