Agarose SFR™ Super Fine Resolution, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
10207
100503-644EA 929.12 USD
100503-644
Agarose SFR™ Super Fine Resolution, Electron Microscopy Sciences
Electrophoresis Gels Agarose and Acrylamide
(C6H10O5·C6H8O4)n CAS 9012-36-6
Agarose SFR™ is a high resolution sieving agarose with unsurpassed clarity. DNA bands differing in size by 2% can be resolved in the range of 200 – 1,000 bp. This agarose is suitable for the analysis of AFLP’s (Amplified Fragment Length Polymorphisms), STR’s (Short Tandem Repeats) and tetranucleotide repeats. The low melting point of agarose SFR™ makes it an excellent medium for analytical and preparative electrophoresis. ™ trademark of Amresco®