MAC X-ray Fluorescence Standards, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
80090-20 80093-CR 80091-10 80093-RH 80093-LA 80093-W 80093-Y 80093-YB 80093-ZN 80093-ZR 80093-CS 80093-CU 80093-DY 80093-EU 80093-FE 80093-GA 80093-GD 80093-GE 80093-HF 80093-HG 80093-HO 80093-I 80093-K 80093-MG 80093-MN 80093-MO 80093-NA 80093-NB 80093-ND 80093-NI 80093-O 80093-PB 80093-PD 80093-PR 80093-PT 80093-RB 80093-S 80093-SB 80093-SE 80093-SI 80093-SM 80093-SN 80093-SR 80093-TA 80093-TE 80093-TH 80093-TI 80093-U 80093-V
100499-688EA 3735.67 USD
100499-688 100503-598 100499-690 100503-602 100503-600 102099-130 102099-132 102099-134 102099-136 102099-138 102099-052 102099-054 102099-056 102099-058 102099-060 102099-062 102099-064 102099-066 102099-068 102099-070 102099-072 102099-074 102099-076 102099-078 102099-080 102099-082 102099-084 102099-086 102099-088 102099-090 102099-092 102099-094 102099-096 102099-098 102099-100 102099-102 102099-104 102099-106 102099-108 102099-110 102099-112 102099-114 102099-116 102099-118 102099-120 102099-122 102099-124 102099-126 102099-128
MAC X-ray Fluorescence Standards, Electron Microscopy Sciences
Standards Color Reference and Spectroscopy Standards
MAC X-ray Microanalysis standards are supplied as 33 m (1/4") diameter pressed pellets or metal foils, and are prepared from selected high purity elements and compounds to ensure interference free-spectra. They are suitable for use by experienced spectroscopists as well as staff training.
Each of the pressed pellets is supported by a thin-walled aluminum cup which is protected from damage during handling and minimizes the risk of touching the standard materials, some of which are harmful
Universal Set of 20 Elements :Na, Mg, Al, Si, P, S, Cl, Ca, Ti, Cr, Mn, Fe, Ni, Cu, Zn, Nb, Sn, Ba, W, Pb
Rare Earth Set of 10 Elements: La, Ce, Pr, Nd, Sm, Eu, Gd, Dy, Ho, Yb
Precious Metal Set of 5 Elements: Rh, Pd, Ag, Pt, Au
The precious metal foils are approximately 0.125 mm thick and are stretched across plastic supports. Standards are available for 60 elements.