RC Filter Flow Air Dryer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74838
100502-812EA 1369.93 USD
100502-812
RC Filter Flow Air Dryer, Electron Microscopy Sciences
Heaters Hot Air Dryers
This RC filter flow air dryer means 5 minutes to perfect flat, and spotless RC prints or films.
A built in air filter eliminates dust. It is thermostatically controlled for optimal drying temperatures. Accommodates four 11×14" prints. Measures 19½×14×7¼" (49.5×35.5×18.5 cm).