Collodion 2% in amyl acetate, non-filter
Supplier: Electron Microscopy Sciences
Highly purified nitrocellulose (parlodion strip) in glass distilled amyl acetate. Useful for forming a negative replica to very fine detail.
Two types are available in range Electron Microscopy Sciences: Sterile formula which is filtered down to 0.45 micron and our non-sterile formula.
We are now offering Collodion (Parlodian) in a 30 ml bottle to avoid its classification as a hazardous chemical for shipping purposes.
MDL Number:
MFCD00081525 CAS Number: 9004-70-0 UN: 2059 ADR: 3,II |