Screwdriver Set, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62079
100502-364EA 37.14 USD
100502-364
Screwdriver Set, Electron Microscopy Sciences
Screwdrivers
This 6-in-1 fold up screwdriver set consists of 6 different screwdriver blades that folds up like a pocket knife and serves as a handle. Blades are 1/8, 3/16 and 1/4" flat. Set also includes T-15 Star Torx, #1 Phillips and Hole Punch awl.