mBox® Microarray Slide Handling, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72188-01
100502-204EA 25.3 USD
100502-204
mBox® Microarray Slide Handling, Electron Microscopy Sciences
Microscope Slide Boxes
mBox® provides microarray handling without removing slides.
  • Immerse 20 slides in liquid during blocking, denaturating, and post-arraying
  • It separates and prevents slide damage during shipping and processing
  • Optimally designed rack spacing for easy slide removal
  • Fits standard microtiter plate formats
  • Manufactured to prevent slide contamination
  • A convenient removable handle available
Melting point: 168 °C (334 °F)