Silicone Staining Pad, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71565
100502-092EA 41.39 USD
100502-092
Silicone Staining Pad, Electron Microscopy Sciences
Staining Tiles
Made from white silicone, a non-reactive material.
  • Pad has 40 cells in 5 rows of 8 each
  • Measures L×W×T: 127×76×13 mm (5×3×½")
Each cell is half-sphere shaped with an opening of 6 mm diameter and 5 mm deep. A few drops of staining solution is added to the wells and grids are then immersed and retrieved as per staining procedure. A watch-glass plate comes with each dish to reduce oxygen and evaporation.