Staining Jar, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70316-04 70316-02
100502-042PK 341 USD
100502-042 100490-750
Staining Jar, Electron Microscopy Sciences
Jars Staining Jars
An all glass jar which holds 5 to 10 standard slides (3 x 1").
They can be used as a chromatography jar with a choice of 2 ground glass lids. The lids are ground to the flange of the jar which deters evaporation. The jar with a taller lid accommodates larger slides: 100 x 25 mm (4 x 1").