Stereo Viewers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68100
100501-592EA 59.45 USD
100501-592 100496-268
Stereo Viewers, Electron Microscopy Sciences
Magnifiers
A simple device, used for viewing a set of 3-dimensional aerial photos in 3-dimension. It comes with two support legs that are folded flat for storage and are unfolded for the viewing position. Photos are viewed through the two lenses simultaneously with both eyes. Lenses may be adjusted for suitable eye-to-eye width. The coated lenses are 32 mm in diameter.
2X Size: 118 x 56 x 124 mm, Net Weight: 227g
4X Size: 118 x 56 x 82 mm, Net Weight: 216g