Measure for Measure, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
31200
100501-574EA 32.05 USD
100501-574
Measure for Measure, Electron Microscopy Sciences
Books
At last, a GREAT Conversion Factor Handbook!
Contains over 39,000 conversions for over 5100 different units.
864 pages
Comprehensive SI reference (the International Standard)
Thousands of foreign and ancient conversion factors
Accurate to 8 significant figures
Value listed in both standard and scientific notation
Handy 4 x 6” size with a durable Lexotone cover