SEM Specimen Preparation Stand, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76750
100501-372EA 132.1 USD
100501-372
SEM Specimen Preparation Stand, Electron Microscopy Sciences
Specimen Mounting Supplies
Specimen mounts of different sizes are placed in the outer ring of the holder base. The center hole of the base will hold a bottle of conductive adhesive. One side of the base takes 10 of the 1/8" pin mounts; reverse side takes 10 of 10 to 15 mm diameter mounts.