Tungsten Wire, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
73805 73804 73801 73800 73802 73803
100501-358EA 230.28 USD
100501-358 100502-118 100492-108 100492-106 100502-116 100501-356
Tungsten Wire, Electron Microscopy Sciences
Wire
We are offering tungsten wire in 99.90% purity.
Available in 0.025" (.64 mm), 0.02" (.51 mm), 0.009" (0.23 mm) diameters.