Magnifier 8x For 35 mm Slide, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68125
100501-332EA 62.16 USD
100501-332
Magnifier 8x For 35 mm Slide, Electron Microscopy Sciences
Magnifiers
Well designed magnifier for high performance.
The 8X magnifier uses a 24 x 37 mm angular objective lens and a two-element achromatic lens for the eyepiece. This arrangement enables the 8X magnifier to cover the entire 24 x 36 mm picture area of a single frame of 35mm film, and it is suitable for checking negatives. Engraved on the bottom edge of the transparent acrylic skirt is a 30 mm long, 1 mm division scale, which does not interfere with the field of view. With all of these features, you should have one for every day use.
Measurements: 46 x 49 x 39 mm.
Weight: 52 g