Aluminium oxide, γ-phase Type N, powder 0.05 µm

Supplier: Electron Microscopy Sciences

50360-01
100501-314LB 122.55 USD
100501-314
Aluminium oxide, γ-phase Type N, powder 0.05 µm
Aluminium oxide, γ-phase

EMS's Alumina Powders are accurately controlled for all specs and particle size distribution, assuring you the best polishing results.


The following micron sizes are available: 0.05, 0.3, and 1.0.


Type DX Alumina Powder is specially treated to reduce the number of agglomerates (group of particles in each grade of manufactured alumina). The result is fast polishing and improved quality of the finished products.
Type N Alumina Powder is recommended when these improvements are not necessary.


Typical applications are:
Precision Optics
Metallographic Sections
Electro-Optical Crystals
Silicone Wafers
Ferrite Components
Gem Stones
Acrylic lenses

Formula: Al₂O₃
MDL Number: MFCD00003424
CAS Number: 1344-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR