Personal Antimicrobial Wipes -P.A.W.S., Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70576-50 70576-01
100501-212PK 26.53 USD
100501-212 100489-190
Personal Antimicrobial Wipes -P.A.W.S., Electron Microscopy Sciences
Wipes Disposable Wipes and Towels Disinfectant Wipes
P.A.W.S. provides sanitizing and extended antimicrobial action on the skin with an individually packaged. Pre-moistened 5×8” towelette. Great for hands, P.A.W.S. is formulated with PCMX (Chloroxylenol -an antimocrobial agent) and Alcohol and provides the most rapid and greatest reduction of microbial counts on the skin (helping you comply with OSHA Bloodborne Pathogen Regulations). In addition, P.A.W.S. contains Aloe Vera to condition the skin and maintain dermal integrity.
Packaging: 100 packets per dispenser box, and 50 towelette in a dispenser container.