Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72652
100500-904BX 145.22 USD
100500-904 100502-646 100500-908 100500-902 100500-906 100500-900
Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
Vials
CYO-LOK vials are ideal for storage or transport of material of extremely low temperatures.
  • Graduated in 0.5 ml increments
The vials have external threads, providing a smooth and uniform inner surface. This eliminates the problem of air born contamination and/or sample loss so common when vials with internal threads are used. The screw top incorporates a molded-in sealing ring, which assures a tight leak-proof seal.