Micro Cover Glasses, Square and Rectangular Sizes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72200-10 72204-03 72204-40 72204-01 72210-10 72204-04 72210-20 72204-20 72200-21 72200-41 72198-10 72200-40 72210-11 72200-30 72204-02 72200-31 72210-21 72198-11 72200-20 72204-10 72204-30
100500-804BX 22.08 USD
100500-804 100500-816 100502-850 100500-812 100500-820 100500-818 100500-822 100502-846 100502-838 100502-842 100500-800 100500-810 100502-852 100500-808 100500-814 100502-840 100502-854 100493-672 100500-806 100502-844 100502-848
Micro Cover Glasses, Square and Rectangular Sizes, Electron Microscopy Sciences
Cover Glasses
The Micro Cover Glasses are manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant.
  • Uniform in size and thickness
  • Square slides are 22 x 22 mm
  • Rectangular slides are 22 mm wide and 30, 40 or 50 mm long