Repetitive Syringe Dispenser, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72484 72487 72483 72486 72482 72480 72489 72485 72488
100500-658BX 227.12 USD
100500-658 100488-476 100500-656 100500-662 100500-654 100491-966 100501-180 100500-660 100501-178
Repetitive Syringe Dispenser, Electron Microscopy Sciences
Syringe Dispensing Systems
Syringe dispenser offers a fast and accurate way of dispensing multiple doses of a set-volume. The volume delivered is determined by the volume-selector-dial and the size of the syringe chosen. A chart comes with the unit to help you to choose the size of syringe, volume, and the number of dispenses. The set dispenses 21 different volumes up to 58 times without refilling, with an accuracy of +/- 1.5% or better depending on the size of the syringe. All syringes are autoclavable, except 60 mL size. Technical Data tests are based on water at a temperature of 25°C.