Five-Slide Gripper, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71410-06
100499-574EA 106.85 USD
100499-574
Five-Slide Gripper, Electron Microscopy Sciences
Racks Staining Racks
Five-slide gripper accomodates microscope slides in one staining procedure. It is made from a special material which is resistant to all chemicals and solvents which are used in staining.
  • No need to remove slides for drying
  • Drying temps up to 80 °C
  • Fits coplin or round-open staining jars