Makler Counting Chamber, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68052-01
100499-400EA 3332.69 USD
100499-400
Makler Counting Chamber, Electron Microscopy Sciences
Slides Counting Chambers
The Makler counting chamber is used for rapid semen analysis. It is only 10 microns deep, which is one tenth the depth of other hemacytometers, making it the shallowest of known chambers.
This quality chamber is constructed from two pieces of optically flat glass: the first is the chamber, the second is the cover glass, which has a fine grid pattern of 1mm squares and a center area further subdivided into 0.1 mm squares. Four quartz pins of precise height hold the cover glass to give an exact trapped specimen depth of 10 microns.