KOMET™ Spinbar® - Stirring Bar, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64517-01
100498-846EA 191.88 USD
100498-846
KOMET™ Spinbar® - Stirring Bar, Electron Microscopy Sciences
Stirrers Magnetic Stirring Bars
The 'PTFE' Komet™ Spinbar® Magnetic Stirring Bar is made from a high-energetic magnetic material.
  • Improved efficiency
  • The Samarium-Cobalt magnet is encapsulated in 'PTFE'
  • The risk of demagnetization caused by external magnetic fields is completely eliminated
Combined with its octagonal shape, produce exceptional power although its length is only 50 mm (1.969") with a 21 mm (0.82") diameter. Its eight edges generate strong turbulence so liquids are thoroughly mixed and its stability is not impaired in vessels with curved bottoms. Komet™ transmits torque loads two to three times larger than those of conventional stirring bars of this length significantly improving efficiency.