VIKEM® Vinyl Coated Lead Wire, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64443-00
100498-824EA 63.55 USD
100498-824
VIKEM® Vinyl Coated Lead Wire, Electron Microscopy Sciences
Wire
This lead wire is ¼" (6.4 mm) in diameter and is coated with a chemical-resistant red Vikem vinyl.
Its 12" (300 mm) length and its flexibility make it a very versatile weight. It can be coiled around a flask, beaker, or any other irregular shape. It weighs only ¼ lb (114 g).