Pipette Work Station, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64385-09
100502-544EA 318.06 USD
100502-544 100498-768
Pipette Work Station, Electron Microscopy Sciences
Pipette Stands
EMS pipette work stations are clear acrylic bins with clear hinged self closing acrylic lid
Bins are available in both single and double unit. The single unit has 4-position holder for pipettors, and the double bin unit has 7 positions.