Pipettor Workstation, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64374-01
100498-734EA 374.63 USD
100498-734
Pipettor Workstation, Electron Microscopy Sciences
Pipette Stands
Allows you to keep the pipettor and the disposable tips handy at all times in one convenient box.
  • Self-closing lid provides dust-free storage
  • Built-in rack stores up to 6 pipettors
  • Bottom, sides, and top pipettor rack are made from white acrylic
  • Bins have a transparent front for easy identification of their contents
  • Two bins, each measure 5"(H) x 8"(W) x 3" (D)
  • The holes at top are 17/32" in diameter
Overall measurements: 9½" (H) x 16 3/8" (W)
Weight 5.8 lbs.