Glass Knife Scoring Jig, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62192-02
100498-010EA 180.15 USD
100498-010
Glass Knife Scoring Jig, Electron Microscopy Sciences
Microtomes and Microtome Assemblies Microtome Blades and Handles
These set of jigs assist in the production of making glass knives by hand. When the edge of the T-square jig is placed against the edge of the glass knife strip, the  subsequent score will be 26 mm (1") long. After breaking, the square of the glass can be scored at a 45 degree angle by using the other jig. 