Diamond Points, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62155-23 62155-38 62155-13 62155-18 62155-52 62155-35 62155-63 62155-83 62155-29 62155-73 62155-24
100497-974EA 37.02 USD
100497-974 100497-982 100497-970 100497-972 100497-984 100497-980 100497-986 100497-990 100497-978 100497-988 100497-976
Diamond Points, Electron Microscopy Sciences
Biopsy Punches
All diamond points are mounted on 2.35 mm (3/32”) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.