Mixing Spatulas, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62127-11 62127-13 62127-12 62127-14
100497-864EA 17.83 USD
100497-864 100497-868 100497-866 100497-870
Mixing Spatulas, Electron Microscopy Sciences
Spatulas
Made from polished stainless steel.
They are ideal for applying wax to glass knives, colloidal adhesives to stubs, and for mixing embedding resins.