Phillips® Screwdriver Set, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62077-15 62077-00 62077-25 62077-20 62077-12
100497-576EA 10.96 USD
100497-576 100497-572 100497-580 100497-578 100497-574
Phillips® Screwdriver Set, Electron Microscopy Sciences
Screwdrivers
A very handy set of Phillips screwdrivers which feature one of the smallest sizes available (1.20 mm). A knurled body with hexagonal heads and tempered steel blades.