Alumina slurry 1 µm

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

50368-30
100497-272EA 49.55 USD
100497-272
Alumina slurry 1 µm
Aluminium oxide

These suspensions are water-based and contain no additives, which may interfere with polishing or contaminate the sample. All slurries are very stable and alumina is evenly dispersed. Excellent for the finest finishes.

Formula: Al₂O₃
MDL Number: MFCD00003424
CAS Number: 1344-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR