Gold, disc, self cleaning foil disc apertures 3 mm O.D., Thickness 0.24 mm

Supplier: Electron Microscopy Sciences

44060-AU 44150-AU 44005-AU 44025-AU 44020-AU 44040-AU 44400-AU 44010-AU 44030-AU 44200-AU 44070-AU 44100-AU 44300-AU 44050-AU 44015-AU
100496-712EA 212.51 USD
100496-712 100496-724 100496-680 100496-696 100496-692 100496-704 100496-736 100496-684 100496-700 100496-728 100496-716 100496-720 100496-732 100496-708 100496-688
Gold, disc, self cleaning foil disc apertures 3 mm O.D., Thickness 0.24 mm
Gold

Advanced techniques of thin metal film application permits the apertures to retain a clean hole edge during prolonged beam exposure


Gold foil aperture tolerances vary as follow:
• Nominal hole size from 1 to 60 micron: +/-1micron
• Nominal hole size 70 micron and up: +/-10%


Ultra-thin, half-micron design discourages contaminant accumulation and stabilizes astigmatic conditions to provide longer life. Conventional mechanical cleaning involving costly down-time is no longer necessary. Because the foil is ultra thin, care must be taken when handling the gold foil apertures.

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
MDL Number: MFCD00003436
CAS Number: 7440-57-5
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR