Gold, disc, self cleaning foil disc apertures 3 mm O.D., Thickness 0.12 mm
Supplier: Electron Microscopy Sciences
Advanced techniques of thin metal film application permits the apertures to retain a clean hole edge during prolonged beam exposure
Gold foil aperture tolerances vary as follow:
• Nominal hole size from 1 to 60 micron: +/-1micron
• Nominal hole size 70 micron and up: +/-10%
Ultra-thin, half-micron design discourages contaminant accumulation and stabilizes astigmatic conditions to provide longer life. Conventional mechanical cleaning involving costly down-time is no longer necessary. Because the foil is ultra thin, care must be taken when handling the gold foil apertures.
Formula:
Au |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...